Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502765007
Encompass_Preferred
The Grainger 16T115 is a 115-volt, 60 Hz, single phase, open drip-proof electric motor with a 1/3 horsepower rating. It has a 1075 RPM operating speed and features a 48 frame size with a C-face mounting. This motor has thermal overload protection, manual reset, and meets NEMA design B specifications. It requires a 230V power supply and is suitable for various industrial applications.
Catalog No. 50-276-5007 Supplier Grainger Supplier No. 16T115
The Grainger 16T115 is a 115-volt, 60 Hz, single phase, open drip-proof electric motor with a 1/3 horsepower rating. It has a 1075 RPM operating speed and features a 48 frame size with a C-face mounting. This motor has thermal overload protection, manual reset, and meets NEMA design B specifications. It requires a 230V power supply and is suitable for various industrial applications.