Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502768560
Encompass_Preferred
The Micro 100 Threading Tool is a 5/16" shank threading tool with a 60 degree threading insert. It has a 0.625" max depth of thread and can cut unified screw threads from 4-56 to 1"-8. The tool body is made from C2 carbide and uses TPG432 inserts. It has a TiN coating for increased tool life. The threading tool has a 5/16" square shank and is 5.5 inches overall. It is designed for general purpose threading applications.
Catalog No. 50-276-8560 Supplier Grainger Supplier No. 16T239
The Micro 100 Threading Tool is a 5/16" shank threading tool with a 60 degree threading insert. It has a 0.625" max depth of thread and can cut unified screw threads from 4-56 to 1"-8. The tool body is made from C2 carbide and uses TPG432 inserts. It has a TiN coating for increased tool life. The threading tool has a 5/16" square shank and is 5.5 inches overall. It is designed for general purpose threading applications.