Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502766473
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 5/16 inch shank diameter. It has a 60 degree threading point and can cut unified coarse, unified fine, and 8 pitch threads. The overall length is 5 inches and it has an American National Standard LH threading direction. This single point threading tool allows quick threading passes on lathes, screw machines, and CNC lathes. It features precision ground threads for accurate, repeatable internal and external threading. The tool is made of M2 high speed steel for strength and durability.
Catalog No. 50-276-6473 Supplier Grainger Supplier No. 16T364
The Micro 100 Threading Tool is a high speed steel threading tool with a 5/16 inch shank diameter. It has a 60 degree threading point and can cut unified coarse, unified fine, and 8 pitch threads. The overall length is 5 inches and it has an American National Standard LH threading direction. This single point threading tool allows quick threading passes on lathes, screw machines, and CNC lathes. It features precision ground threads for accurate, repeatable internal and external threading. The tool is made of M2 high speed steel for strength and durability.