Learn More
Grainger Threading Tool 5/16 Carbide
Supplier:  Grainger 16T366
Encompass_Preferred
The Grainger Approved Threading Tool is a high-speed steel threading tool with a 60 degree threading angle. It has a 1/2 inch shank diameter and is 6 inches overall length. The insert size is 3/8 inch with 16 threads per inch. This threading tool is designed for right-hand threads and has a TiN coating for increased tool life. It can thread a variety of materials including steel, stainless steel, cast iron, aluminum and plastics. The Grainger Approved brand offers reliable tools at a value price point. This threading tool would be a good option for cutting quality right-hand threads on lathes and screw machines.
Catalog No. 50-276-6805