Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502767313
Encompass_Preferred
The Micro 100 Threading Tool is a high-speed steel threading tool with a 5/16" shank diameter and 16 TPI. It has a 60 degree included angle and can cut up to 5/8" depth of thread in steel, stainless steel, cast iron, aluminum, and other materials. The tool has an AlTiN nano coating for increased tool life. It is made in the USA by Micro 100, a manufacturer of indexable tooling, solid carbide tools, and other precision cutting tools. This threading tool allows efficient internal and external threading operations on CNC lathes and screw machines.
Catalog No. 50-276-7313 Supplier Grainger Supplier No. 16T118
The Micro 100 Threading Tool is a high-speed steel threading tool with a 5/16" shank diameter and 16 TPI. It has a 60 degree included angle and can cut up to 5/8" depth of thread in steel, stainless steel, cast iron, aluminum, and other materials. The tool has an AlTiN nano coating for increased tool life. It is made in the USA by Micro 100, a manufacturer of indexable tooling, solid carbide tools, and other precision cutting tools. This threading tool allows efficient internal and external threading operations on CNC lathes and screw machines.