Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502767350
Encompass_Preferred
The Micro 100 Threading Tool is a high-speed steel threading tool with a 5/16" shank diameter and 60 degree threading point. It has an overall length of 3-1/2" and can cut unified coarse right-hand threads from 4-48 to 1"-8 TPI. The tool features TiN coating for increased tool life and has an included angle of 60 degrees. It is designed for use on lathes, screw machines, and CNC lathes when cutting unified coarse right-hand internal threads. The threading tool has a screw-lock style holder and is made in the USA.
Catalog No. 50-276-7350 Supplier Grainger Supplier No. 16T177
The Micro 100 Threading Tool is a high-speed steel threading tool with a 5/16" shank diameter and 60 degree threading point. It has an overall length of 3-1/2" and can cut unified coarse right-hand threads from 4-48 to 1"-8 TPI. The tool features TiN coating for increased tool life and has an included angle of 60 degrees. It is designed for use on lathes, screw machines, and CNC lathes when cutting unified coarse right-hand internal threads. The threading tool has a screw-lock style holder and is made in the USA.