Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502767380
Encompass_Preferred
The Grainger 5/16"-18 carbide straight flute machine tap is made of solid carbide for durability and has straight flutes for efficient chip removal. It is designed for hand or machine tapping of ferrous metals like steel, cast iron, and alloy steel. This tap features a standard plug chamfer for easy starting and can tap blind or through holes. It has a spiral point tip to align the tap and reduce wandering. The tap has a bright finish for visibility and comes in a 3" overall length.
Catalog No. 50-276-7380 Supplier Grainger Supplier No. 16T180
The Grainger 5/16"-18 carbide straight flute machine tap is made of solid carbide for durability and has straight flutes for efficient chip removal. It is designed for hand or machine tapping of ferrous metals like steel, cast iron, and alloy steel. This tap features a standard plug chamfer for easy starting and can tap blind or through holes. It has a spiral point tip to align the tap and reduce wandering. The tap has a bright finish for visibility and comes in a 3" overall length.