Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502765410
Encompass_Preferred
The Micro 100 Threading Tool is a 5/16" shank threading tool with a 60 degree threading insert. It has a 0.625" max depth of thread and can cut unified screw threads from 4-56 to 1"-8. The tool body is made from C2 carbide and uses TPG432 inserts. It has a TiN coating for increased tool life. The threading tool has a 5/16" square shank and is 5.5 inches overall. It is designed for general purpose threading applications in most materials. The tool comes with one TPG432 insert. Additional inserts are available for purchase separately.
Catalog No. 50-276-5410 Supplier Grainger Supplier No. 16T175
The Micro 100 Threading Tool is a 5/16" shank threading tool with a 60 degree threading insert. It has a 0.625" max depth of thread and can cut unified screw threads from 4-56 to 1"-8. The tool body is made from C2 carbide and uses TPG432 inserts. It has a TiN coating for increased tool life. The threading tool has a 5/16" square shank and is 5.5 inches overall. It is designed for general purpose threading applications in most materials. The tool comes with one TPG432 insert. Additional inserts are available for purchase separately.