Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502765631
Encompass_Preferred
The Grainger 5/16"-18 carbide straight flute machine tap is made of solid carbide for durability and has straight flutes for efficient chip removal. It is designed for hand or machine tapping of ferrous metals like steel, cast iron, and alloy steel. This tap features a standard plug chamfer and is suitable for blind holes. It has a spiral point tip to guide the tap and reduce thrust and friction. The tap has a bright finish for visibility and comes in a 3" length that can be used by hand or in a machine.
Catalog No. 50-276-5631 Supplier Grainger Supplier No. 16T181
The Grainger 5/16"-18 carbide straight flute machine tap is made of solid carbide for durability and has straight flutes for efficient chip removal. It is designed for hand or machine tapping of ferrous metals like steel, cast iron, and alloy steel. This tap features a standard plug chamfer and is suitable for blind holes. It has a spiral point tip to guide the tap and reduce thrust and friction. The tap has a bright finish for visibility and comes in a 3" length that can be used by hand or in a machine.