Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502768555
Encompass_Preferred
The Scientific Cutting Tools 5/16-24 Port Thread Port Tool is a handheld tool used for deburring and chamfering holes. It has a 5/16-24 threaded shank and is made of high speed steel for durability. The port tool features a 60 degree included angle and is designed for use on steel, stainless steel, aluminum, brass, bronze, and more. It provides an economical solution for deburring, beveling, and chamfering holes up to 1/2" diameter. The tool has an overall length of 3-1/2" and weighs 0.05 lbs.
Catalog No. 50-276-8555 Supplier Grainger Supplier No. 16T249
The Scientific Cutting Tools 5/16-24 Port Thread Port Tool is a handheld tool used for deburring and chamfering holes. It has a 5/16-24 threaded shank and is made of high speed steel for durability. The port tool features a 60 degree included angle and is designed for use on steel, stainless steel, aluminum, brass, bronze, and more. It provides an economical solution for deburring, beveling, and chamfering holes up to 1/2" diameter. The tool has an overall length of 3-1/2" and weighs 0.05 lbs.