Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502766884
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 5/16 inch shank diameter and 1/2 inch maximum depth of thread. It has a 60 degree threading point angle and can cut unified coarse, unified fine, and 8 pitch threads. The overall tool length is 3-1/2 inches. This single point threading tool allows quick threading starts and smooth thread finishes in most materials.
Catalog No. 50-276-6884 Supplier Grainger Supplier No. 16T360
The Micro 100 Threading Tool is a high speed steel threading tool with a 5/16 inch shank diameter and 1/2 inch maximum depth of thread. It has a 60 degree threading point angle and can cut unified coarse, unified fine, and 8 pitch threads. The overall tool length is 3-1/2 inches. This single point threading tool allows quick threading starts and smooth thread finishes in most materials.