Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502767272
Encompass_Preferred
The Grainger 16T117 is a 16 ton hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/4 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It has a two piece handle for easy pumping and controlled lifting. The jack's capacity is 16 tons (32,000 lbs). It weighs 43 pounds and has a one year warranty. The jack is designed for garage, auto repair, machinery and plant maintenance applications.
Catalog No. 50-276-7272 Supplier Grainger Supplier No. 16T117
The Grainger 16T117 is a 16 ton hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/4 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It has a two piece handle for easy pumping and controlled lifting. The jack's capacity is 16 tons (32,000 lbs). It weighs 43 pounds and has a one year warranty. The jack is designed for garage, auto repair, machinery and plant maintenance applications.