Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502768598
Encompass_Preferred
The Micro 100 Threading Tool is a high-speed steel threading tool with a 5/16" shank diameter and 60 degree included angle. It has a 1/4" minimum minor diameter and can cut up to 8 threads per inch. The overall tool length is 3-1/2". This single-point threading tool allows both external and internal threading operations. It features a TiN coating for increased tool life and lubricity. The Micro 100 threading tool is made in the USA.
Catalog No. 50-276-8598 Supplier Grainger Supplier No. 16T369
The Micro 100 Threading Tool is a high-speed steel threading tool with a 5/16" shank diameter and 60 degree included angle. It has a 1/4" minimum minor diameter and can cut up to 8 threads per inch. The overall tool length is 3-1/2". This single-point threading tool allows both external and internal threading operations. It features a TiN coating for increased tool life and lubricity. The Micro 100 threading tool is made in the USA.