Learn More
Grainger Threading Tool 5/16 Carbide
Catalog No. 502764004
Encompass_Preferred
The Grainger 5/16"-18 carbide straight flute machine tap is made of solid carbide for durability and has straight flutes for efficient chip removal. It is designed for hand or machine tapping of ferrous metals like steel, cast iron, and alloy steel. The tap has a spiral point for easy starting and cutting edges with high wear resistance. It comes in a standard length of 2-1/2" with a square shank and is rated for moderate cutting speeds and feeds. Overall, this is a high quality carbide tap well-suited for tapping common sized threaded holes in hard metals.
Catalog No. 50-276-4004 Supplier Grainger Supplier No. 16T067
The Grainger 5/16"-18 carbide straight flute machine tap is made of solid carbide for durability and has straight flutes for efficient chip removal. It is designed for hand or machine tapping of ferrous metals like steel, cast iron, and alloy steel. The tap has a spiral point for easy starting and cutting edges with high wear resistance. It comes in a standard length of 2-1/2" with a square shank and is rated for moderate cutting speeds and feeds. Overall, this is a high quality carbide tap well-suited for tapping common sized threaded holes in hard metals.