Learn More
Grainger Threading Tool 5/16 Carbide
Supplier:  Grainger 16T050
Encompass_Preferred
The Scientific Cutting Tools 5/16" Shank Threading Tool is a high speed steel threading tool with a 5/16" shank diameter and 60 degree threading point. It has an overall length of 5-1/2" and can cut unified coarse right hand threads from 4 to 56 threads per inch. The tool features a TiN coating for increased tool life and lubricity. It is made in the USA by Scientific Cutting Tools, a manufacturer specializing in high performance cutting tools since 1955. This threading tool allows efficient threading operations on lathes, screw machines, and CNC machines.
Catalog No. 50-276-3735