Learn More
Grainger Threading Tool 6.00mm Carbide
Catalog No. 502767699
Encompass_Preferred
The Micro 100 Threading Tool is a high-speed steel threading tool with a 6mm shank diameter and 16mm maximum cutting diameter. It has a 60 degree threading angle and can cut metric, UN, and UNF threads. The overall length is 3.5 inches. This single-point threading tool allows quick threading passes with superior chip control. It is designed for use on lathes and screw machines. The tool comes with a TiN coating for increased tool life and lubricity. It is made in the USA by Micro 100, a leading manufacturer of indexable tooling, solid carbide drills, solid carbide end mills, and other precision cutting tools.
Catalog No. 50-276-7699 Supplier Grainger Supplier No. 16T195
The Micro 100 Threading Tool is a high-speed steel threading tool with a 6mm shank diameter and 16mm maximum cutting diameter. It has a 60 degree threading angle and can cut metric, UN, and UNF threads. The overall length is 3.5 inches. This single-point threading tool allows quick threading passes with superior chip control. It is designed for use on lathes and screw machines. The tool comes with a TiN coating for increased tool life and lubricity. It is made in the USA by Micro 100, a leading manufacturer of indexable tooling, solid carbide drills, solid carbide end mills, and other precision cutting tools.