Learn More
Grainger Threading Tool 6.00mm Carbide
Catalog No. 502768299
Encompass_Preferred
The Micro 100 Threading Tool is a high-speed steel threading tool with a 6mm shank diameter and 16mm maximum cutting diameter. It has a 60 degree threading profile and can cut metric, UN, and UNF threads. The overall tool length is 3 inches. This single-point threading tool allows both external and internal threading operations. It features TiN coating for increased tool life. The Micro 100 threading tool is made in the USA.
Catalog No. 50-276-8299 Supplier Grainger Supplier No. 16T199
The Micro 100 Threading Tool is a high-speed steel threading tool with a 6mm shank diameter and 16mm maximum cutting diameter. It has a 60 degree threading profile and can cut metric, UN, and UNF threads. The overall tool length is 3 inches. This single-point threading tool allows both external and internal threading operations. It features TiN coating for increased tool life. The Micro 100 threading tool is made in the USA.