Learn More
Grainger Threading Tool 6.00mm Carbide
Catalog No. 502765951
Encompass_Preferred
The Micro 100 Threading Tool is a high-performance threading tool with a 6mm shank diameter and 16mm maximum cutting diameter. It has a TiN coating for increased tool life and can thread a variety of materials including steel, stainless steel, cast iron, aluminum, brass, bronze, plastics and more. The tool features precision ground threads for accurate threading and includes a screw-lock style holder. It is made in the USA out of Micro-100's proprietary carbide grade to deliver consistent tool life and performance. This is an ideal threading tool for difficult to machine materials.
Catalog No. 50-276-5951 Supplier Grainger Supplier No. 16T202
The Micro 100 Threading Tool is a high-performance threading tool with a 6mm shank diameter and 16mm maximum cutting diameter. It has a TiN coating for increased tool life and can thread a variety of materials including steel, stainless steel, cast iron, aluminum, brass, bronze, plastics and more. The tool features precision ground threads for accurate threading and includes a screw-lock style holder. It is made in the USA out of Micro-100's proprietary carbide grade to deliver consistent tool life and performance. This is an ideal threading tool for difficult to machine materials.