Learn More
Grainger Threading Tool 8.00mm Carbide
Catalog No. 502768625
Encompass_Preferred
The Micro 100 Threading Tool is a high-speed steel threading tool with an 8mm shank diameter and 16mm maximum cutting diameter. It has a 60 degree threading angle and can cut metric, UNC, UNF, and BA thread forms. The overall tool length is 3.5 inches. This single-point threading tool allows both external and internal threading operations. It features TiN coating for increased tool life and lubricity. The Micro 100 threading tool is made in the USA.
Catalog No. 50-276-8625 Supplier Grainger Supplier No. 16T215
The Micro 100 Threading Tool is a high-speed steel threading tool with an 8mm shank diameter and 16mm maximum cutting diameter. It has a 60 degree threading angle and can cut metric, UNC, UNF, and BA thread forms. The overall tool length is 3.5 inches. This single-point threading tool allows both external and internal threading operations. It features TiN coating for increased tool life and lubricity. The Micro 100 threading tool is made in the USA.