Learn More
Grainger Threading Tool 8.00mm Carbide
Catalog No. 502766020
Encompass_Preferred
The Threading Tool 8.00mm Carbide 16T208 is a high speed steel threading tool with an 8mm shank diameter and 16mm maximum depth of thread. It has a 60 degree threading angle and features TiAlN coating for increased tool life and performance. The tool is made of M2 high speed steel and has an overall length of 208mm. It can be used on lathes for both internal and external threading applications on materials like steel, stainless steel, cast iron and non-ferrous metals. The unique design allows for easy chip removal during threading operations. Overall, this is a durable and versatile threading tool for 8mm applications.
Catalog No. 50-276-6020 Supplier Grainger Supplier No. 16T208
The Threading Tool 8.00mm Carbide 16T208 is a high speed steel threading tool with an 8mm shank diameter and 16mm maximum depth of thread. It has a 60 degree threading angle and features TiAlN coating for increased tool life and performance. The tool is made of M2 high speed steel and has an overall length of 208mm. It can be used on lathes for both internal and external threading applications on materials like steel, stainless steel, cast iron and non-ferrous metals. The unique design allows for easy chip removal during threading operations. Overall, this is a durable and versatile threading tool for 8mm applications.