Learn More
Grainger Threading Tool 8.00mm Carbide
Catalog No. 502768336
Encompass_Preferred
The Micro 100 Threading Tool is a high-performance threading tool with an 8mm shank diameter and 16mm maximum cutting diameter. It has a TiN coating for increased tool life and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, and more. The tool features a precision ground thread profile to ensure accurate, repeatable threading. With its rigid design, this threading tool is well-suited for production threading applications. The tool comes with a 30 degree entering angle.
Catalog No. 50-276-8336 Supplier Grainger Supplier No. 16T210
The Micro 100 Threading Tool is a high-performance threading tool with an 8mm shank diameter and 16mm maximum cutting diameter. It has a TiN coating for increased tool life and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, and more. The tool features a precision ground thread profile to ensure accurate, repeatable threading. With its rigid design, this threading tool is well-suited for production threading applications. The tool comes with a 30 degree entering angle.