Learn More
Narishige International Usa Inc Narishige MMO-203 Three-Axis Hydraulic Micromanipulator, Fine Movement, Vibration-Free Control
Catalog No. NC3468757
Encompass
The Narishige MMO-203 is a three-axis hydraulic micromanipulator that enables precise, vibration-free micro-manipulation for scientific applications. It features excellent linear control and smooth movements, with remote operation and compatibility with various pipette holders. Recommended for cell-holding in microinjection systems, it stands out due to its cost-efficient design compared to joystick methods, and its adaptability for both right and left-handed use enhances ergonomic comfort.
Catalog No. NC3468757 Supplier Narishige International Usa Inc Supplier No. MMO203
Add to Cart
Add to Cart
Request a Quote
The Narishige MMO-203 is a three-axis hydraulic micromanipulator that enables precise, vibration-free micro-manipulation for scientific applications. It features excellent linear control and smooth movements, with remote operation and compatibility with various pipette holders. Recommended for cell-holding in microinjection systems, it stands out due to its cost-efficient design compared to joystick methods, and its adaptability for both right and left-handed use enhances ergonomic comfort.