Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Three Bear Family™ Sort, Pattern, and Play Activity Set
SCI_ED

Catalog No. S79391
Change view
Click to view available options
Grade:
Pre-K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S79391 Pre-K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

CHOKING HAZARD —Small parts. Not for children under 3 yrs.

Catalog No. S79391 Supplier Learning Resources, Inc Supplier No. LER 0757
Only null left
Add to Cart
Add to Cart

CHOKING HAZARD —Small parts. Not for children under 3 yrs.

Develop early math skills with a variety of colorful bear manipulatives.

Set includes 96 Three Bear Family counters in six colors, six sorting bowls, 2 in. color cube, number cube, 16 pattern cards, 12 double-sided activity cards and bear game spinner. Also includes a colorful plastic storage tub with carrying handle and removable sorting tray. Grades PK-2.

Specifications

Grade Level Early Childhood, Elementary, Intermediate - Middle School, High School
Grade Pre-K to 12
Format Hands-on Learning
Product Type Three Bear Family™ Sort, Pattern and Play Activity Set
Includes 96 Three bear family counters in six colors, six sorting bowls, 2 in. color cube, number cube, 16 pattern cards, 12 double sided activity cards and bear game spinner
For Use With (Application) Visual learning methods for better understanding and performance
Subject Mathematics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.