Learn More
S-Curve™ Three Compartment Glove Dispenser with Individual Lids, Front Access
Catalog No. 19999430 Shop All S-Curve Technologies Products
Change view
Click to view available options
Material:
Acrylic
PETG
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-999-430 Acrylic
19-162-424 PETG
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-999-430 Supplier S-Curve™ Supplier No. GD300
Add to Cart
Add to Cart
This item is not returnable. View return policy
Three compartment dispenser includes three individually opening lids, with front access holes for easy product dispensing.
  • Three compartment dispenser with individually opening lids
  • Front access holes for easy product dispensing
  • Can be mounted with heavy-duty wall brackets (included)
  • Flame treated edges for a smooth finish
Specifications
Product Type Glove Dispenser
Color Clear
Dimensions (D x W x H) Interior 12 x 16 x 12 in.
Holds Gloves
Material Acrylic
Mounting Keyhole Slots
No. of Compartments 3
Product Style Top Loading
Includes Heavy-duty wall mounting bracket
Thickness (English) 0.25 in.