Learn More
Thomas Scientific Traceabler 3-Channel Jumbo Timer
SDP
Catalog No. 501461549
Encompass
3 channels, plus memory Resolution is 1 second Accuracy is 0.01% Timing capacity: 99 hours, 59 minutes, 59 seconds to 1 second Clock is provided with an individually serial numbered Traceable® certificate from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST. Count-up/count-down/clock includes easel stand and magnet. Size is 2” x 3 1/4” x 1/2”. Weight is 3 oz. Please note: The aforementioned A2LA accreditation belongs to the manufacturer, not Thomas Scientific.
Catalog No. 50-146-1549 Supplier Thomas Scientific Supplier No. 1235D46
3 channels, plus memory Resolution is 1 second Accuracy is 0.01% Timing capacity: 99 hours, 59 minutes, 59 seconds to 1 second Clock is provided with an individually serial numbered Traceable® certificate from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST. Count-up/count-down/clock includes easel stand and magnet. Size is 2” x 3 1/4” x 1/2”. Weight is 3 oz. Please note: The aforementioned A2LA accreditation belongs to the manufacturer, not Thomas Scientific.