Learn More
Marketlab Three Mode Handheld Metal Detector
Description
- Three modes of operation allow for high-performance ferrous only detection around the head and body or full metal detection around the body.
- Capable of separately screening for both ferrous and nonferrous metallic items.
- Eliminates risks of dislocation, torsion, overheating, or induction of electrical current.
- Audible, silent-vibration, and visual-magnitude alert settings.
- Visual signaling combined with the audio provide feedback on the extent of the alarm.
- Personalize the interface, energy-saving mode, and sensitivity of the three operational modes.
- Refine the analysis capabilities based on specific patient needs.
- Rechargeable battery can be used for up to 24 hours between charges.
- Comes with a charger and carry case.
Specifications
Specifications
| Languages | English |
| Product Type | Handheld Metal Detector |