Learn More
3B Scientific™ Three-Part Female Pelvis with Ligaments - includes 3B Smart Anatomy
GREENER_CHOICESDPSCI_ED
Catalog No. S96431 Shop All 3B Scientific Products
Change view
Click to view available options
Includes:
With Ligaments
Without Ligaments
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S96431 Without Ligaments
S96430 With Ligaments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S96431 Supplier 3B Scientific™ Supplier No. H20/1
Add to Cart
Add to Cart
Represents an original cast of a bony female pelvis, which shows all details of anatomical structures
3B Smart Anatomy is now included with any 3B Scientific anatomy model.
Every original 3B Scientific anatomy model now includes these additional FREE features:
  • Free access to the anatomy course 3B Smart Anatomy, hosted inside the award-winning Complete Anatomy app by 3D4Medical
  • The 3B Smart Anatomy course includes 23 digital anatomy lectures, 117 different virtual anatomy models and 39 anatomy quizzes to test your knowledge
  • FREE warranty upgrade from 3 to 5 years with every product registration
TIPP: you will also receive access to a free 3-day trial to all premium features of the Complete Anatomy app when you sign up for your 3B Smart Anatomy course.
To unlock these benefits, simply scan the label located on your model and register online. All 3B Smart Anatomy features are completely free of charge.
  • Shows two hip bones, the pubic symphysis, the sacrum, and the coccyx, the fifth lumbar vertebra with intervertebral disc
  • Midsagital section through the fifth lumbar vertebra, sacrum and coccyx, allow both halves of pelvis to disassembles
  • Left half of fifth lumbar vertebra is removable
Ligamented Model
  • Right half shows following pelvic ligaments: inguinal, sacrotuberous, sacrospinous, anterior sacroiliac, iliolumbar, anterior longitudinal, interosseous sacroiliac, posterior sacroiliac, and obturator membrane
Specifications
Includes Without Ligaments
Weight (English) 1.98 lb.
Weight (Metric) 0.9 kg
Product Type Model, Pelvis, 3-Part Female