Learn More
Chemglass Life Sciences Clamp, Three-Prong Swivel
SDP
Catalog No. CG921501
Encompass_Preferred
Clamp, Three-Prong Swivel. Dual adjustment clamp with shaft wing nut to allow clamp jaw movement of 360°. Rod holder grips up to 3/4” (19mm) diameter. The clamp has a 2 1/4” (57mm) maximum grip size with a 5/8” (16mm) O.D. extension arm. Complete with removable vinyl and fiberglass covers.
Catalog No. CG921501 Supplier Chemglass Life Sciences Supplier No. CG921501
Clamp, Three-Prong Swivel. Dual adjustment clamp with shaft wing nut to allow clamp jaw movement of 360°. Rod holder grips up to 3/4” (19mm) diameter. The clamp has a 2 1/4” (57mm) maximum grip size with a 5/8” (16mm) O.D. extension arm. Complete with removable vinyl and fiberglass covers.