Learn More
Electron Microscopy Sciences Three Prong Tweezers, 4 3/4" Long
SDP
Catalog No. 5029314
Encompass_Preferred
This device is actuated by one thumb. Its three prong claw is handy in the picking up of small items up to 2 1/4" in diameter.
Catalog No. 50-293-14 Supplier Electron Microscopy Sciences Supplier No. 72905
This device is actuated by one thumb. Its three prong claw is handy in the picking up of small items up to 2 1/4" in diameter.