Learn More
BURKLE INC Three-way valve, PP/PE, Ø 0.20-0.28 in. (5-7 mm), NW 0.16 in. (4 mm), red/white
Catalog No. 5000025619
Encompass_Preferred
Three-way valves are used for transfer of liquids in order to release the flow, to block it or to change its direction.
Catalog No. 50-000-25619 Supplier BURKLE INC Supplier No. 86060060
Three-way valves are used for transfer of liquids in order to release the flow, to block it or to change its direction.