Learn More
BURKLE INC Three-way valve, PP/PE, Ø 0.35-0.43 in. (9-11 mm), NW 0.31 in. (8 mm), red/white
Catalog No. 5000025760
Encompass_Preferred
Three-way valves are used for transfer of liquids in order to release the flow, to block it or to change its direction.
Catalog No. 50-000-25760 Supplier BURKLE INC Supplier No. 86060100
Three-way valves are used for transfer of liquids in order to release the flow, to block it or to change its direction.