Learn More
BURKLE INC Three-way valve, PVDF, Ø 0.28-0.35 in. (7-9 mm), NW 0.24 in. (6 mm), white
Catalog No. 5000025858
Encompass_Preferred
Three-way valves are used for transfer of liquids in order to release the flow, to block it or to change its direction.
Catalog No. 50-000-25858 Supplier BURKLE INC Supplier No. 86080080
Three-way valves are used for transfer of liquids in order to release the flow, to block it or to change its direction.