Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Three60™ Sampling System
GREENER_CHOICE

Catalog No. 4MP0039 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1 L
2 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
4MP0039 2 L
4MP0034 50 mL
4MP0035 100 mL
4MP0036 250 mL
4MP0037 500 mL
4MP0038 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 4MP0039 Supplier Thermo Scientific™ Supplier No. 4MP0039
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allows you to take multiple representative samples of your process liquids conveniently and in a closed system.

The Thermo Scientific™ Three60™ Sampling System allows you to take a representative sample of your product with minimal effort. The fluid paths of the four pre-irradiated BioProcess Containers (BPCs) and assemblies feature quick-turn valves and pinch-and-cut disconnections for fluid transfer and removal. The Three60 Sampling System is composed of Thermo Scientific™ ASI™ 77 film, a 5-layer, 7mL, polyethylene film and includes 2 ports (1 luer lock insert + cap, 1 injection site).

  • Compatible with a 3.8 cm (1.5-inch) sanitary connector
  • Requires no expensive hardware
  • Each Three60 package contains a valve and four assemblies with pinch-and-cut disconnections
  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks

Applications

  • Removes the need for tank cleaning and helps to reduce cycle times
  • Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2 L
Content And Storage Store at room temperature.
Dimensions 35.0 x 19.0 cm (13.78 x 7.48 in.)
Film Description ASI™ 77
Outer Packaging Placed into a thermoform tray and sealed
Port Detail 2 Ports (1 Luer Lock Insert + Cap, 1 Injection Site)
Material Ethyl Vinyl Acetate
Quantity 8/Cs.
Shipping Condition Room Temperature
Sterilization Method Irradiated
Product Line Three60
Product Type Sampling System
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.