Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

threo-5,6-Dodecanediol 96.0+%, TCI America™
SDP

Catalog No. D26781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26781G Supplier TCI America Supplier No. D26781G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 70859-33-5
Molecular Formula C12H26O2
Molecular Weight (g/mol) 202.338
MDL Number MFCD02093463
InChI Key COLYTQSCHUMMSR-RYUDHWBXSA-N
Synonym threo-5,6-Dihydroxydodecane
PubChem CID 90476545
IUPAC Name (5S,6S)-dodecane-5,6-diol
SMILES CCCCCCC(C(CCCC)O)O

Specifications

Melting Point 47°C
Color White
Quantity 1 g
Formula Weight 202.34
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material threo-5,6-Dodecanediol
EINECSNumber (2)-0240
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.