Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Threo-9,10-dihydroxystearic acid | 2391-05-1 | MFCD00046726 | 99.6% | 316.48 g/mol | C18H36O4 | 10 MG
SDP

Catalog No. 5000284039
Encompass_Preferred

Threo-9,10-dihydroxystearic acid is a synthetic dihydroxy fatty acid used in biochemical research to probe lipid bilayer packing and membrane fluidity. It is supplied as a solid reagent for laboratory studies and analytical applications.

  • Synthetic dihydroxy fatty acid used to study membrane packing and fluidity.
  • Chemical formula C18H36O4 and molecular weight 316.48 g/mol.
  • CAS number 2391-05-1 for unambiguous identification.
  • Available in small research quantities: 1 mg, 5 mg, 10 mg.
  • Store at 4°C, protect from light, and store under inert gas for long-term stability.
  • Consult the certificate of analysis for lot-specific purity and analytical data.

Catalog No. 50-002-84039 Supplier Medchemexpress LLC Supplier No. HYW70043710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Threo-9,10-dihydroxystearic acid is a synthetic dihydroxy fatty acid used in biochemical research to probe lipid bilayer packing and membrane fluidity. It is supplied as a solid reagent for laboratory studies and analytical applications.

  • Synthetic dihydroxy fatty acid used to study membrane packing and fluidity.
  • Chemical formula C18H36O4 and molecular weight 316.48 g/mol.
  • CAS number 2391-05-1 for unambiguous identification.
  • Available in small research quantities: 1 mg, 5 mg, 10 mg.
  • Store at 4°C, protect from light, and store under inert gas for long-term stability.
  • Consult the certificate of analysis for lot-specific purity and analytical data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.