Learn More
Medchemexpress LLC Threo-9,10-dihydroxystearic acid | 2391-05-1 | MFCD00046726 | 99.6% | 316.48 g/mol | C18H36O4 | 25 MG
SDP
Catalog No. 5000284538
Encompass_Preferred
Threo-9,10-dihydroxystearic acid is a synthetic dihydroxy fatty acid used in research to modify lipid bilayer packing and membrane fluidity. It is supplied as a solid in small laboratory quantities for biochemical and biophysical studies.
  • Alters lipid bilayer packing and membrane fluidity.
  • Synthetic dihydroxy fatty acid with formula C18H36O4.
  • High purity appropriate for research applications.
  • Available in small laboratory quantities such as 25 mg.
  • White to off-white solid, suitable for standard laboratory storage.
Catalog No. 50-002-84538 Supplier Medchemexpress LLC Supplier No. HYW70043725MG
Threo-9,10-dihydroxystearic acid is a synthetic dihydroxy fatty acid used in research to modify lipid bilayer packing and membrane fluidity. It is supplied as a solid in small laboratory quantities for biochemical and biophysical studies.
  • Alters lipid bilayer packing and membrane fluidity.
  • Synthetic dihydroxy fatty acid with formula C18H36O4.
  • High purity appropriate for research applications.
  • Available in small laboratory quantities such as 25 mg.
  • White to off-white solid, suitable for standard laboratory storage.