Learn More
Medchemexpress LLC Threo-9,10-dihydroxystearic acid | 2391-05-1 | MFCD00079162 | 99.6% | 316.48 | C18H36O4 | 100 MG
SDP
Catalog No. 5000284620
Encompass_Preferred
Threo-9,10-dihydroxystearic acid is a synthetic dihydroxy fatty acid used as a biochemical assay reagent to probe lipid bilayer packing and membrane fluidity. Supplied for research use only, it is suitable for membrane biophysics studies, model membranes, and lipid-protein interaction assays.
  • Synthetic dihydroxy fatty acid for membrane studies.
  • Modulates lipid bilayer packing and fluidity.
  • Suitable for biochemical assays and model membrane systems.
  • Provided with supporting analytical data (COA, HPLC, MS) where available.
  • Store cold and protected from light to preserve stability.
Catalog No. 50-002-84620 Supplier Medchemexpress LLC Supplier No. HYW700437100MG
Threo-9,10-dihydroxystearic acid is a synthetic dihydroxy fatty acid used as a biochemical assay reagent to probe lipid bilayer packing and membrane fluidity. Supplied for research use only, it is suitable for membrane biophysics studies, model membranes, and lipid-protein interaction assays.
  • Synthetic dihydroxy fatty acid for membrane studies.
  • Modulates lipid bilayer packing and fluidity.
  • Suitable for biochemical assays and model membrane systems.
  • Provided with supporting analytical data (COA, HPLC, MS) where available.
  • Store cold and protected from light to preserve stability.