Learn More
Medchemexpress LLC Threo-dihydrobupropion (hydrochloride) | 357637-18-4 | 99.0% | 278.22 | C13H21Cl2NO | 5 MG
SDP
Catalog No. 5000263988
Encompass_Preferred
Threo-dihydrobupropion hydrochloride is the hydrochloride salt of threo-dihydrobupropion, a primary metabolite of the antidepressant bupropion. It is supplied as a solid research chemical intended for use as a reference standard and for in vitro or in vivo studies of depression-related pharmacology, behavioral assays, and metabolic or biochemical investigations.
  • Primary metabolite of bupropion.
  • Hydrochloride salt supplied as a solid for laboratory use.
  • Suitable for depression, behavioral, and biochemical research applications.
  • Available in small-mass packages for analytical and pharmacological assays.
  • Characterized by CAS 357637-18-4 and molecular weight 278.22.
Catalog No. 50-002-63988 Supplier Medchemexpress LLC Supplier No. HY1314795MG
Threo-dihydrobupropion hydrochloride is the hydrochloride salt of threo-dihydrobupropion, a primary metabolite of the antidepressant bupropion. It is supplied as a solid research chemical intended for use as a reference standard and for in vitro or in vivo studies of depression-related pharmacology, behavioral assays, and metabolic or biochemical investigations.
  • Primary metabolite of bupropion.
  • Hydrochloride salt supplied as a solid for laboratory use.
  • Suitable for depression, behavioral, and biochemical research applications.
  • Available in small-mass packages for analytical and pharmacological assays.
  • Characterized by CAS 357637-18-4 and molecular weight 278.22.