Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

threo Ifenprodil hemitartrate, Tocris Bioscience™

Catalog No. 289210
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-9210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 28-9210 Supplier R&D Systems Supplier No. 2892/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

GluN2B-selective NMDA antagonist; also σ agonist

Potent σ receptor agonist (Ki values are 59.1 and 2 nM for σ1 and σ2 receptors respectively) and GluN2B (formally NR2B) subunit-selective NMDA receptor antagonist (IC50 values are 0.22 and 324.8 μM at GluN2B and GluN2A respectively). Displays ∽8-fold reduced affinity at α-adrenoceptors compared to Ifenprodil (Cat.No. 0545). Inhibits the hERG potassium channel (IC50 = 88 nM) and exhibits antiarrhythmic activity in vivo.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Chemical Identifiers

CAS 1312991-83-5
Molecular Formula C92H120N4O20
Molecular Weight (g/mol) 1601.98
InChI Key MMUFFGLAKFWLGJ-RRQZTXAJSA-N
Synonym threo ifenprodil hemitartrate
PubChem CID 90488783
IUPAC Name 4-[(1S,2S)-2-(4-benzylpiperidin-1-yl)-1-hydroxypropyl]phenol;4-[(1R,2R)-2-(4-benzylpiperidin-1-yl)-1-hydroxypropyl]phenol;2,3-dihydroxybutanedioic acid
SMILES CC(C(C1=CC=C(C=C1)O)O)N2CCC(CC2)CC3=CC=CC=C3.CC(C(C1=CC=C(C=C1)O)O)N2CCC(CC2)CC3=CC=CC=C3.CC(C(C1=CC=C(C=C1)O)O)N2CCC(CC2)CC3=CC=CC=C3.CC(C(C1=CC=C(C=C1)O)O)N2CCC(CC2)CC3=CC=CC=C3.C(C(C(=O)O)O)(C(=O)O)O.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Quantity 10 mg
Formula Weight 400.49
Percent Purity >98%
Chemical Name or Material threo Ifenprodil hemitartrate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.