Learn More
MSC FLIR ONE Pro Thermal Camera, Smartphone, -20-400 C, 1 ea
Catalog No. 501909454
Encompass
The FLIR ONE Pro-Series camera helps users find invisible problems faster, whether inspecting electrical panels, troubleshooting mechanical systems, looking for HVAC issues, or finding water damage. It offers enhanced image clarity with VividIR™ and and detects temperature differences down to 70 mK, providing powerful measurement tools.
  • MSX image enhancement for detailed thermal pictures.
  • VividIR™ image processing for sharper clarity.
  • Measures temperatures up to 400°C (752°F).
  • Drop-tested to withstand 1.8 meters.
  • Connects to smart devices via the app for live feed and cloud upload.
  • Thermal resolution of 160 x 120 (native).
Catalog No. 50-190-9454 Supplier MSC Supplier No. 49276330
The FLIR ONE Pro-Series camera helps users find invisible problems faster, whether inspecting electrical panels, troubleshooting mechanical systems, looking for HVAC issues, or finding water damage. It offers enhanced image clarity with VividIR™ and and detects temperature differences down to 70 mK, providing powerful measurement tools.
  • MSX image enhancement for detailed thermal pictures.
  • VividIR™ image processing for sharper clarity.
  • Measures temperatures up to 400°C (752°F).
  • Drop-tested to withstand 1.8 meters.
  • Connects to smart devices via the app for live feed and cloud upload.
  • Thermal resolution of 160 x 120 (native).