Learn More
Andwin Scientific THRML CMBO CRYOTAGS 500/PK
SDP
Supplier:  Andwin Scientific 91386000
Encompass_Preferred
The Direct Thermal Combo Cryo-Tags can be utilized in liquid nitrogen and within the temperature range of -196C to 70C. Using Dymo printers, print the amount you need, and put the labels on the desired item. The labels measure 1.5" L x 0.5" W and have a diameter of 3/8". There are 400 labels in a pack.
Catalog No. NC0314228