Learn More
Sigma Aldrich Fine Chemicals Biosciences Thrombin Active High Activ
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SRP6556100KU
Encompass
Thrombin is a coagulation protein and a serine protease (EC 3.4.21.5) that catalyzes many coagulation-related reactions.
Catalog No. 50-291-4245