Learn More
Cusabio Technology LLC thrombin-antithrombin complex
Catalog No. 5000049102
Encompass_Preferred
This kit detects TAT (Human thrombin-antithrombin complex) in serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a broad detection range of 62 5 pg/mL to 4000 pg/mL with high sensitivity reliably measuring concentrations as low as 15 6 pg/mL The assay requires a sample volume of 50-100 L per test and delivers results within 1 to 5 hours Measurements are performed at an absorbance wavelength of 450 nm ensuring precise quantification of TAT levels for research or diagnostic applications related to coagulation disorders thrombotic events or hemostasis studies The kit includes optimized reagents and protocols for consistent performance across biological matrices
Catalog No. 50-000-49102 Supplier Cusabio Technology LLC Supplier No. CSBE08431H
This kit detects TAT (Human thrombin-antithrombin complex) in serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a broad detection range of 62 5 pg/mL to 4000 pg/mL with high sensitivity reliably measuring concentrations as low as 15 6 pg/mL The assay requires a sample volume of 50-100 L per test and delivers results within 1 to 5 hours Measurements are performed at an absorbance wavelength of 450 nm ensuring precise quantification of TAT levels for research or diagnostic applications related to coagulation disorders thrombotic events or hemostasis studies The kit includes optimized reagents and protocols for consistent performance across biological matrices