Learn More
Cusabio Technology LLC thrombin-antithrombin complex
Catalog No. 5000050827
Encompass_Preferred
This kit detects TAT (thrombin-antithrombin complex) in pig serum plasma and tissue homogenates using a quantitative sandwich ELISA method It provides a detection range of 62 5 pg/mL to 4000 pg/mL with a sensitivity of 15 6 pg/mL enabling precise measurement of TAT levels The assay requires 50-100 L of sample per test and involves an incubation period of 1-5 hours Absorbance is measured at 450 nm for accurate quantification making it suitable for research applications requiring reliable analysis of thrombin-antithrombin complex concentrations in various biological matrices
Catalog No. 50-000-50827 Supplier Cusabio Technology LLC Supplier No. CSBE13995P
This kit detects TAT (thrombin-antithrombin complex) in pig serum plasma and tissue homogenates using a quantitative sandwich ELISA method It provides a detection range of 62 5 pg/mL to 4000 pg/mL with a sensitivity of 15 6 pg/mL enabling precise measurement of TAT levels The assay requires 50-100 L of sample per test and involves an incubation period of 1-5 hours Absorbance is measured at 450 nm for accurate quantification making it suitable for research applications requiring reliable analysis of thrombin-antithrombin complex concentrations in various biological matrices