Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Thrombin, Bovine

Catalog No. 08820361 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 kU/Pk., 5 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-820-361 2 kU/Pk., 5 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-820-361 Supplier MP Biomedicals™ Supplier No. 08820361
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thrombin, Bovine is the final coagulation protease in regard to hemostasis, promoting both procoagulant and anticoagulant effects.

  • Thrombin is used for site specific cleavage of recombinant fusion proteins containing an accessible thrombin recognition site for removal of affinity tags.
  • Thrombin has been used in a study to assess in vitro hemostatic properties of French lyophilized plasma.
TRUSTED_SUSTAINABILITY

Specifications

pH Range 6.8
Form Powder
Quantity 2 kU/Pk., 5 Pk.
Synonym Factor Ila

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.