Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombin Bovine High Purity Grade MP Biomedicals

Catalog No. icn15416301 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15416301 1 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15416301 Supplier MP Biomedicals, Inc Supplier No. 0215416301
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12-4-9002
Molecular Formula C18H26N5O14P
Molecular Weight (g/mol) 567.40
InChI Key VPXDHZMLJOJKOX-UHFFFAOYNA-N
Synonym uricase, urate oxidase, urate:o2 oxidoreductase, uricase, candida utilis, uricase from candida sp., urate: oxygen oxoreductase, uricase from bacillus fastidiosus, uricase from arthrobacter globiformis, upu, uricase from bacillus fastidiosus, lyophilized, ∼9 u/mg
PubChem CID 14228836
IUPAC Name azane;[5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl [5-(2,4-dioxopyrimidin-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl] hydrogen phosphate
SMILES C1=CN(C(=O)NC1=O)C2C(C(C(O2)COP(=O)(O)OC3C(OC(C3O)N4C=CC(=O)NC4=O)CO)O)O.N

Specifications

Assay Percent Range 10%
Quantity 1 kU
Grade High Purity
Chemical Name or Material Thrombin Bovine, From Bovine Plasma Activity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.