Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Thrombin, Bovine Plasma

Catalog No. p-5386246
Change view
Click to view available options
Quantity:
100KU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63383EXP 100KU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63383EXP Supplier Thermo Scientific Chemicals Supplier No. J63383EXP
Only null left
Add to Cart

For research use

Thrombin is a trypsin-like serine protease involved in a multitude of processes in the human body. Thrombin generation is the result of limited proteolysis of the vitamin K-dependent zymogen prothrombin. Thrombin is the last enzyme in the clotting cascade functioning to cleave fibrinogen to fibrin which forms the fibrin gel of a haemostatic plug or a pathologic thrombus

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized powder, Reconstitute in 0.9% sodium chloride
Quantity 100KU
Source Bovine plasma
Storage Requirements -30° to -10°C
Regulatory Status RUO
Product Type Thrombin, Bovine Plasma
Recombinant Native
Form Lyophilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.