Learn More
Sigma Aldrich Fine Chemicals Biosciences Thrombin CleanCleaveTM Kit
Catalog No. 502906481
Encompass
The Thrombin CleanCleave Kit contains a 50% suspension fo thrombin-agarose produced by immobilizing bovine thrombin and is designed for cleavage of recombinant fusion proteins
Catalog No. 50-290-6481 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. RECOMT1KT
The Thrombin CleanCleave Kit contains a 50% suspension fo thrombin-agarose produced by immobilizing bovine thrombin and is designed for cleavage of recombinant fusion proteins