Learn More
Sigma Aldrich Fine Chemicals Biosciences Thrombin from human plasma
Catalog No. 502901120
Encompass
Thrombin is a coagulation factor IIa. It is a serine endopeptidase that hydrolyzes peptide and ester bonds specifically at the carboxylic side of Arg. The enzyme converts fibrinogen to fibrin. The product contains EDTA and additional stabilizing agents.
Catalog No. 50-290-1120 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10602400001
Thrombin is a coagulation factor IIa. It is a serine endopeptidase that hydrolyzes peptide and ester bonds specifically at the carboxylic side of Arg. The enzyme converts fibrinogen to fibrin. The product contains EDTA and additional stabilizing agents.