Learn More
Sigma Aldrich Fine Chemicals Biosciences Thrombin human BioUltra re
Catalog No. 502904421
Encompass
Thrombin is the final coagulation protease in regard to hemostasis promoting both procoagulant and anticoagulant effects.
Catalog No. 50-290-4421 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T9326150UN
Thrombin is the final coagulation protease in regard to hemostasis promoting both procoagulant and anticoagulant effects.